BP5 GC Capillary Column
Actual product may vary
Thermo Scientific
BP5 GC Capillary Column
A general purpose phase used for a wide range of applications
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric) InnerFilm Thickness
SGE05416030 m0.53 mm3 μm
Catalog number SGE054160
Price (MYR)
4,825.00
Each
Length (Metric):
30 m
Diameter (Metric) Inner:
0.53 mm
Film Thickness:
3 μm
Price (MYR)
4,825.00
Each
  • Excellent general purpose column
  • Low bleed
  • Non-polar
  • High temperature
  • Ideal for aromatics, pesticides, herbicides, drugs of abuse and hydrocarbons
Specifications
For Use With (Application)Phenols Analysis, Ultra Trace Analyses, Pesticides and Herbicides Analysis, Hydrocarbons, Solvents Analysis, Phenols, Amines, GC-MS and Other Specific Detector Applications
Film Thickness3 μm
Length (Metric)30 m
Packing MaterialForte™ BPX5
PolarityNon polar
CoatingFused Silica
USP TypeG27, G36, G41
Stationary Phase5% Phenyl Polysilphenylene Siloxane
Diameter (Metric) Inner0.53 mm
Functionality/Interaction TypePi Bonding with the Aromatic Phenyl Groups
Max. Temperature (Metric)350/360°C
Temperature Range (Metric)-40°C to +350/360°C
Unit SizeEach