Potassium Chloride, Certified AR for Analysis, Fisher Chemical™
Potassium Chloride, Certified AR for Analysis, Fisher Chemical™
Potassium Chloride, Certified AR for Analysis, Fisher Chemical™
Fisher Chemicals
Potassium Chloride, Certified AR for Analysis, Fisher Chemical™
ClK, CAS Number-7447-40-7, klotrix, muriate of potash, potassium chloride kcl, chlorvescent, kalitabs, klor-con, enseal, potassium chloride, sylvite, slow-k, 500g, 1420 deg.C, CHEBI:32588, White, 74.54g/mol, WCUXLLCKKVVCTQ-UHFFFAOYSA-M, potassium;chloride, 770 deg.C, 11360, 74.548
Have Questions?
Catalog NumberQuantityPackaging
P428060
also known as P/4280/60
1 kgPlastic powder jar
Catalog number P428060
also known as P/4280/60
HSN 28273990
Price (MYR)
-
Quantity:
1 kg
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
Boiling Point1420°C
Melting Point770°C
CAS7447-40-7
ColorWhite
pH6
View more
ClK, CAS Number-7447-40-7, klotrix, muriate of potash, potassium chloride kcl, chlorvescent, kalitabs, klor-con, enseal, potassium chloride, sylvite, slow-k, 500g, 1420 deg.C, CHEBI:32588, White, 74.54g/mol, WCUXLLCKKVVCTQ-UHFFFAOYSA-M, potassium;chloride, 770 deg.C, 11360, 74.548