Navigator™ NV2201
Navigator™ NV2201
Actual product may vary
Ohaus
Navigator™ NV2201
Offers accuracy and repeatability at a competitive price point, making it a valuable tool in lab, industrial and education settings. OHAUS™ Navigator™ NV2201 is designed for a range of essential weighing applications.
Have Questions?
Catalog number OHA30467933
Price (MYR)
399.00
EA
Add to cart
Price (MYR)
399.00
EA
Add to cart
  • Designed to accurately stabilize in one second to provide fast and accurate weighing results
  • Overload protection system that can withstand loads up to four times the rated capacity safeguards the balance
  • An integrated shipping lock protects the weighing cell during transportation/storage
  • Rugged
Specifications
Certifications/ComplianceCAN/CSA, WEEE, RoHS, UL
Connection TypeAC Adapter
Capacity2200 g
Display TypeLCD
Readability0.1 g
IncludesAC Adapter
MaterialABS Housing, Stainless Steel Pan
ModelNV2201
Stabilization Time1 sec.
TypeAnalytical Balance
Length (Metric)230 mm
Width (Metric)204 mm
Height (Metric)70 mm
Weight (Metric)0.87 kg
Electrical Requirements120 V
Unit SizeEA