Cleanroom Vinyl Tape, 38 mm (1.5 in.)
Cleanroom Vinyl Tape, 38 mm (1.5 in.)
Actual product may vary
Micronova
Cleanroom Vinyl Tape, 38 mm (1.5 in.)
This vinyl cleanroom tape with rubber, clean-peel adhesive is extremely conformable, making it ideal for color coding, marking and sealing curved and irregular surfaces.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
MICCR100PC-15WHWhite
FIS19-130-1254White
Catalog number MICCR100PC-15WH
Price (MYR)
380.00
CS
Add to cart
Color:
White
Price (MYR)
380.00
CS
Add to cart
The Micronova Cleanroom Vinyl Tape offers a clean peel from wafer cartridges and cleanroom work surfaces and the matte finish can be written on with a cleanroom pen. Special processing and packaging techniques ensure low particles and extractable ion counts for applications in and around the cleanroom.
  • Vinyl tape with synthetic rubber adhesive
  • Clean peel from wafer cartridges and cleanroom work surfaces
  • Ideal for taping and sealing curved and irregular surfaces
  • Matte finish can be written on with a cleanroom pen
  • Specially processed and packaged for the cleanroom
  • Available in a variety of colors and widths
  • Individually packaged in a polyethylene bag
  • 36 yards wound on a 3 in. plastic core
Specifications
AutoclavableNot Autoclavable
Adhesive TypeRubber
CoatingVinyl
ColorWhite
FinishMatte
FormatRoll
MaterialVinyl, Synthetic Rubber
Product TypeCleanroom Tape
SterilityNon-sterile
For Use With (Application)Sealing, Color Coding, Labeling, Batching and Food Processing Applications
Dimensions (LxW)33 m x 38.1 mm (36 yd. x 1.5 in.)
Length (English)36 yd.
Length (Metric)33 m
Width (English)1.5 in.
Width (Metric)38.1 mm
Unit SizeCS