Micronova™ Autoclave Indicator Tape
Micronova™ Autoclave Indicator Tape
Actual product may vary
Micronova
Micronova™ Autoclave Indicator Tape
Confirms a completed sterilisation cycle in both gravity discharge and vacuum autoclaves. DuPont™ Micronova™ Autoclave Indicator Tape adheres on contact and stays in place through live steam pressure. After use, the tape removes quickly and completely, leaving minimal residue or staining.
Have Questions?
Catalog NumberColor
MICAUTOPC-1Natural, Green, Black
Catalog number MICAUTOPC-1
Price (MYR)
1,084.00
CS
Add to cart
Color:
Natural, Green, Black
Price (MYR)
1,084.00
CS
Add to cart
Confirms a completed sterilisation cycle in both gravity discharge and vacuum autoclaves. DuPont™ Micronova™ Autoclave Indicator Tape adheres on contact and stays in place through live steam pressure. After use, the tape removes quickly and completely, leaving minimal residue or staining.
  • Natural background colour, with printed green indicator lines, changing to black when proper levels of steam pressure and temperature have been achieved.
  • Uniquely printed lines cannot be rubbed off and will not contaminate instruments.
  • Saturated crepe paper backing
  • Synthetic polyisoprene adhesive
  • Lead-free ink indicator
  • Standard sterilisation cycle (with pre-vacuum cycle): 121°C (250°F) for 20 minutes, 132° to 134°C (270 to 274°F) for 4 minutes
  • 25mm x 55m length, wound onto a plastic core.
Specifications
Adhesive TypePolyisoprene, Synthetic
ColorNatural, Green, Black
FormatTape
MaterialCrepe Paper
SterilitySterile
For Use With (Equipment)Gravity discharge and vacuum autoclaves
Quantity1 Ea.
Dimensions (LxW)55 cm x 25.4 cm
Length (English)2.16 in.
Length (Metric)55 mm
Width (English)1 in.
Width (Metric)25.4 mm
Unit SizeCS