▸
Flasks
▸
Erlenmeyer Flasks
▸
Assistent™ DURAN™ Wide Neck Graduated Erlenmeyer Flasks
Actual product may vary
Lovibond
Assistent™ DURAN™ Wide Neck Graduated Erlenmeyer Flasks
For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Karl Hecht™ Assistent™ DURAN™ Wide Neck Graduated Erlenmeyer Flasks are manufactured from durable DURAN borosilicate 3.3 glass.
Have Questions?
Catalog Number
Capacity (Metric)
LOV194150
50 mL
Catalog number
LOV194150
Price (MYR)
2,863.35
Each
Add to cart
Capacity (Metric):
50 mL
Price (MYR)
2,863.35
Each
Add to cart
Acc. to DIN 12385
Available in a range of capacities
Wide neck allows for ease of access to contents
Specifications
Certifications/Compliance
DIN 12385
Diameter (Metric) Outer Neck
34 mm
Capacity (Metric)
50 mL
Material
Borosilicate Glass, Duran Glass
Height (Metric)
85 mm
Mouth Style
Wide
Diameter (Metric) Outer
51 mm