FreeZone™ Bulk Tray
FreeZone™ Bulk Tray
Actual product may vary
Labconco
FreeZone™ Bulk Tray
For FreeZone Bulk Tray Dryers and/or Stoppering Tray Dryers
Have Questions?
Catalog NumberFor Use With (Equipment)
LBC7756200Freezone™ Bulk or Stoppering Tray Dryer
Catalog number LBC7756200
Price (MYR)
0.00
EA
Add to cart
For Use With (Equipment):
Freezone™ Bulk or Stoppering Tray Dryer
Price (MYR)
0.00
EA
Add to cart
For FreeZone Bulk Tray Dryers and/or Stoppering Tray Dryers
  • Can be used with both Stoppering and Bulk Tray Dryers
Specifications
TypeBulk Tray
No. of Shelves0
MaterialStainless Steel
For Use With (Equipment)Freezone™ Bulk or Stoppering Tray Dryer
Width (English)11.5 in.
Width (Metric)29.2 cm
Height (English)0.88 in.
Height (Metric)2.2 cm
Length (English)15 in.
Length (Metric)38.1 cm
Weight (English)3 lb.
Weight (Metric)1.4 kg
Unit SizeEA