▸
Freeze Dryers
▸
Freeze Drying Chambers and Equipment
▸
Glass Trap Insert for CentriVap™ System
Actual product may vary
Labconco
Glass Trap Insert for CentriVap™ System
Protects CentriVap Cold Trap reservoir and lid from corrosives.
Have Questions?
Catalog Number
Capacity (Metric)
LBC7397605
2550 mL
Catalog number
LBC7397605
Price (MYR)
9,978.00
Each
Add to cart
Capacity (Metric):
2550 mL
Price (MYR)
9,978.00
Each
Add to cart
Protects CentriVap Cold Trap reservoir and lid from corrosives.
Included with CentriVap Acid Systems; accessory for other systems
Not compatible with pre-2005 Cold Traps
Specifications
Type
Trap Insert
Capacity (Metric)
2550 mL
No. of Shelves
0
Material
Glass
For Use With (Equipment)
CentriVap™ concentrator cold trap
Weight (English)
4 lb.
Weight (Metric)
1.8 kg