Kimtech™ Purple Nitrile™ Gloves
Actual product may vary
Ansell
Kimtech™ Purple Nitrile™ Gloves
Purple nitrile glove offers maximum protection for higher-risk procedures. Unique color differentiates from latex and other synthetic gloves.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC55083Large
KLC55084X-Large
Catalog number KLC55083
Price (MYR)
0.00
Case
Size:
Large
Price (MYR)
0.00
Case
Purple nitrile glove offers maximum protection for higher-risk procedures. Unique color differentiates from latex and other synthetic gloves.
  • Textured fingertips offers dexterity and tactile sensitivity
  • Latex-free
  • Ambidextrous
  • 9.5 in. length
  • Beaded cuff prevent roll-down and allow for easy donning
  • Powder free
Specifications
Certifications/ComplianceASTM D6978-05 (non-sterile only), Meets USP <800> guidelines
Chemotherapeutic RatedYes
ChlorinatedYes
ApplicationsLaboratory, Healthcare
AQL0.65
ColorPurple
DisposableSingle-use
Exterior FinishPartially Textured
Finger Thickness (English)6 mil
Finger Thickness (Metric)0.15 mm
Latex FreeYes
MaterialNitrile
Palm Thickness (English)4.7 mil
Palm Thickness (Metric)0.12 mm
PowderedPowder Free
Product TypeNitrile Examination Glove
SizeLarge
SterilityNon-sterile
Thickness (English)6 mil
Thickness (Metric)0.15 mm
FDA 510 (k)Yes
Glove Cuff StyleBeaded
Model55083
Quantity100 Gloves/Box
SeriesKimtech™
Tensile Strength21 MPa
Length (English)9.5 in.
Length (Metric)24.13 cm
Unit SizeCase