Lavender™ Nitrile Gloves
Lavender™ Nitrile Gloves
Actual product may vary
Ansell
Lavender™ Nitrile Gloves
Lavender nitrile gloves feature enhanced dexterity with textured fingertips and are suited for a wide variety of tasks.
Have Questions?
Catalog NumberSize
KLC52818Medium
Catalog number KLC52818
Price (MYR)
11,965.00
CS
Add to cart
Size:
Medium
Price (MYR)
11,965.00
CS
Add to cart
Lavender nitrile gloves feature enhanced dexterity with textured fingertips and are suited for a wide variety of tasks.
  • Lavender Nitrile - 9.5 in. length
  • Powder free
  • Ambidextrous
  • Beaded cuffs
  • Non-sterile
  • Textured fingertips
  • Easy glove dispensing
Specifications
Certifications/ComplianceASTM D5151
Chemotherapeutic RatedNo
ApplicationsHealthcare, Laboratory
AQL1.5
ColorPurple
DisposableSingle-use
Exterior FinishPartially Textured
Finger Thickness (English)2.8 mil
Finger Thickness (Metric)0.07 mm
Latex FreeYes
MaterialNitrile
Palm Thickness (English)2 mil
Palm Thickness (Metric)0.05 mm
PowderedPowder Free
SizeMedium
Thickness (English)2.8 mil
Thickness (Metric)0.07 mm
FDA 510 (k)Yes
Model52818
Tensile Strength24 MPa
Length (English)9.5 in.
Length (Metric)24.13 cm
Unit SizeCS