Check Order Status
FlexiPump Tubings
Actual product may vary
Interscience
FlexiPump Tubings
Tubings for use with FlexiPump.
Have Questions?
Catalog Number
Diameter (Metric) Inner
ISC561302
3.2 mm
Catalog number
ISC561302
Price (MYR)
432.60
Pack
Add to cart
Diameter (Metric) Inner:
3.2 mm
Price (MYR)
432.60
Pack
Add to cart
Set of 6 Tygon™ tubings for pump part
Available with inner diameter of 1.6 mm, 3.2 mm, 6.4 mm, or 8 mm
Specifications
Autoclavable
Autoclavable
Diameter (English) Inner
0.12 in.
Diameter (Metric) Inner
3.2 mm
Material
Tygon A-60-F
Thickness (English)
0.12 in.
For Use With
FlexiPump
Sterility
Non-sterile
Color
Clear
Product Type
Tubing