Grant Optima™ Digital Thermostat
Grant Optima™ Digital Thermostat
Actual product may vary
Grant
Grant Optima™ Digital Thermostat
General purpose digital heated circulator provides excellent temperature stability and control for demanding applications requiring accurate temperature control.
Have Questions?
Catalog NumberTemperature RangeExternal Circulation
GRAT1000°C to 100°CNo
Catalog number GRAT100
Price (MYR)
10,675.35
Each
Add to cart
Temperature Range:
0°C to 100°C
External Circulation:
No
Price (MYR)
10,675.35
Each
Add to cart
General purpose digital heated circulator provides excellent temperature stability and control for demanding applications requiring accurate temperature control.
  • Stability ±0.05°C.
  • Clear, bright 4 digit LED display
  • Simple, intuitive user interface: dial and two function buttons.
  • Model available with/without clamp (TClamp)
  • Low-liquid detection (float switch)
  • Fixed over temperature
  • 2 point user calibration
  • Visual alarm
  • TC120 model includes Integral pump for external circulation
Specifications
Controller Type4-digit LED
Capacity (Metric)50 L
CommunicationsDigital
Temperature Range0°C to 100°C
External CirculationNo
Electrical Requirements120 V 1.5 kw 50/60 Hz, 230 V 1.4 kw 50/60 Hz
Voltage120/230 V