QB Series Dry Block Heating Systems, Precision Digital
QB Series Dry Block Heating Systems, Precision Digital
Actual product may vary
QB Series Dry Block Heating Systems, Precision Digital
QB Series Dry Block Heating Systems, Precision Digital
Grant
QB Series Dry Block Heating Systems, Precision Digital
Provides excellent temperature control and a wide choice of interchangeable blocks for maximum flexibility. Grant Instruments™ QB Series Dry Block Heating Systems, Precision Digital combines superb temperature control and uniformity with high quality design and great versatility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Blocks
GRAQBD44
FKS114709582
Catalog number GRAQBD4
Price (MYR)
12,046.65
Each
Add to cart
No. of Blocks:
4
Price (MYR)
12,046.65
Each
Add to cart
Provides excellent temperature control and a wide choice of interchangeable blocks for maximum flexibility. Grant Instruments™ QB Series Dry Block Heating Systems, Precision Digital combines superb temperature control and uniformity with high quality design and great versatility.
  • High power heater for fast heat-up - 25°C to 100°C in only 15 minutes.
  • Simple-to-use rotor plus two keys provide access to the interactive interface for fast, accurate set-up and use
  • Clear digital display - easy to read from a distance for instant reassurance
  • Convenient timer facility, with audible buzzer, for reaction timing and function timing, e.g. delayed heater switch-on/turn-off
  • Wide range of interchangeable blocks enables you to mix and match sample tubes as desired - extraction tool supplied as standard for easy and safe removal of blocks.
  • Optional safety cover - protects samples from contamination and users from accidental contact with hot blocks
  • Overtemperature cut-out protects your samples and your workplace
  • High quality, robust construction in streamlined coolwall aluminum and chemical-resistant plastic - durable in demanding environments
  • Compact footprint to optimise use of benchspace
Specifications
Chamber MaterialAluminum
MaterialChemical Resistant Plastic
Temperature Range (Metric)Ambient +5° to 130°C
TypeDry Bath Block
No. of Blocks4
Temperature Stability0.1° at 37°C
Temperature Uniformity0.1° at 37°C
Timer1 to 999 mins
Length (Metric)390 mm
Width (Metric)200 mm
Height (Metric)100 mm
Depth (Metric)380 mm
Voltage120 or 230 V
Wattage600 w
Unit SizeEach