Interchangeable Block for QB Series Dry Block Heaters
Interchangeable Block for QB Series Dry Block Heaters
Actual product may vary
Grant
Interchangeable Block for QB Series Dry Block Heaters
For use with 0.2mL tubes, strips and 96-well microtitre plates used in molecular biology and biotechnology applications. Grant Instruments™ Interchangeable Block for QB Series Dry Block Heaters can withstand temperatures up to 100°C.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTube SizeNo. of PlacesDepth (Metric)
GRAQB-17H17mm1075 mm hole depth
GRAQB-1313mm1250 mm hole depth
FKS1143091816mm1250 mm hole depth
GRAQB-1010mm2450 mm hole depth
GRAQB-1212mm2450 mm hole depth
Catalog number GRAQB-17H
Price (MYR)
1,739.85
Each
Add to cart
Tube Size:
17mm
No. of Places:
10
Depth (Metric):
75 mm hole depth
Price (MYR)
1,739.85
Each
Add to cart
  • For use with QBD1, QBD2, QBD4, QBH2, QBA1 and QBA2 dry block heating system
  • Accommodates a variety of test tube sizes
  • Available in plain block format without holes
Specifications
For Use With (Equipment)Falcon tubes
MaterialAluminum
No. of Places10
TypeBlock
Tube Size17mm
Dimensions (L x W x H)140 x 50 x 63 mm
Depth (Metric)75 mm hole depth
Unit SizeEach