OLS Aqua Pro Orbital and Linear Shaking Bath
OLS Aqua Pro Orbital and Linear Shaking Bath
Actual product may vary
OLS Aqua Pro Orbital and Linear Shaking Bath
OLS Aqua Pro Orbital and Linear Shaking Bath
OLS Aqua Pro Orbital and Linear Shaking Bath
OLS Aqua Pro Orbital and Linear Shaking Bath
Grant
OLS Aqua Pro Orbital and Linear Shaking Bath
Patented, combined orbital and linear shaking mechanism of the OLS26 allows optimisation of aeration and shear forces mixing, for reproducible results.
Have Questions?
Catalog NumberOrbit
GRAOLS269 mm
Catalog number GRAOLS26
Price (MYR)
969.00
EA
Add to cart
Orbit:
9 mm
Price (MYR)
969.00
EA
Add to cart
Patented, combined orbital and linear shaking mechanism of the OLS26 allows optimisation of aeration and shear forces mixing, for reproducible results.
  • Ambient +5°C to 99°C
  • Operate down to 0°C with accessory cooling
  • Timer and presets
  • Drain tap and transparent lid
  • 120V & 230V models available
  • Adjustable shaking intensity and speed up to 200rpm
  • Universal tray and polycarbonate lid included as standard
  • OLS26 combines adjustable orbital and linear shaking in one bath
  • Excellent temperature performance with stability and uniformity at ±0.1°C
  • Dual point user calibration for optimum accuracy
  • Easy-to-change between orbital and linear motion, simply rotate carrier tray 180 degrees
  • Discrete shaking mechanism maximises working area
  • Easy to insert and remove shaking trays even when loaded
  • Wide range of accessory shaking trays for secure hold on most commonly used vessels
  • Dual display showing temperature and shaking speed
  • Adjustable shaking intensity and speed
  • User-defined high temperature alarm and cut-off
  • Advanced run dry/dry start protection
  • Countdown timer from 1 to 999 minutes
Specifications
Orbit9 mm
Speed Range20 to 200 rpm
Temperature Range (Metric)Ambient +5° to 99°C (0°C to 99°C with accessory cooling)
Holds26 L
Product TypeOrbital/Linear Shaking Bath
Temperature Range (English)41°F to 210.2°F
Temperature Resolution±0.1°C
Temperature UniformityAt 70°C ±°C 0.1
Depth (Metric)565 mm
Height (Metric)325 mm
Width (Metric)335 mm
ModelGrant Instruments OLS Aqua Pro Orbital
IncludesShaking Bath, Universal Shaking Tray, Polycarbonate Lid
Unit SizeEA