Single Scale Graduated Cylinders
Single Scale Graduated Cylinders
Actual product may vary
Fisherbrand
Single Scale Graduated Cylinders
With raised numerals for quick, accurate reading.
Have Questions?
Catalog NumberCapacity (Metric)Description
FKS153599521000 mL1000 mL
Catalog number FKS15359952
Price (MYR)
208.00
EA
Add to cart
Capacity (Metric):
1000 mL
Description:
1000 mL
Price (MYR)
208.00
EA
Add to cart
  • Flared top rims and large spouts facilitate pouring
  • Octagonal shaped bases prevent tipping and rolling
  • Non-wetting polypropylene plastic surface eliminates concave meniscus; makes reading easier and more accurate
  • Autoclavable at 121°C (250°F)
Specifications
Capacity (Metric)1000 mL
MaterialPolypropylene
No. per Pack1
Description1000 mL
Diameter (English)2.6 in.
ScaleSingle
Quantity1 Ea.
Graduations10 mL
AutoclavableAutoclavable
Subdivision10 mL
ShapeRound
Diameter (English) Inside2.4 in.
Base TypeOctagonal
Height (English)17.7 in.
Height (Metric)449.5 mm
TypeGraduated Cylinder
Unit SizeEA