PTFE Micro Magnetic Stir Bar
Actual product may vary
PTFE Micro Magnetic Stir Bar
PTFE Micro Magnetic Stir Bar
Fisherbrand
PTFE Micro Magnetic Stir Bar
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)
FKS118889527 mm2 mm
Catalog number FKS11888952
Price (MYR)
67.00
Pack
Length (Metric):
7 mm
Diameter (Metric):
2 mm
Price (MYR)
67.00
Pack
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
  • Supplied in reusable snap cap vials.
  • For stirring in test tubes, small size beakers, or other containers.
  • Autoclavable.
Specifications
Diameter (Metric)2 mm
Length (Metric)7 mm
TypeStir Bar, Micro
Unit SizePack