Polypropylene Graduated Specimen Containers
Polypropylene Graduated Specimen Containers
Actual product may vary
Fisherbrand
Polypropylene Graduated Specimen Containers
Ideal for specimen collection, transportation and storage
Have Questions?
Catalog NumberCapacity (Metric)Capacity (English)SterilityClosure Included
FKS11799378237 mL8 oz.SterileYes
Catalog number FKS11799378
Price (MYR)
219.00
PK
Add to cart
Capacity (Metric):
237 mL
Capacity (English):
8 oz.
Sterility:
Sterile
Closure Included:
Yes
Price (MYR)
219.00
PK
Add to cart
  • Autoclavable
  • Graduated
  • With or without nonautoclavable polyethylene snap lid
Specifications
Closure IncludedYes
GraduatedYes
MaterialPolypropylene
SterilitySterile
Capacity (English)8 oz.
Capacity (Metric)237 mL
Unit SizePK