Microstirrer Magnetic Stirrer
Microstirrer Magnetic Stirrer
Actual product may vary
Microstirrer Magnetic Stirrer
Microstirrer Magnetic Stirrer
Microstirrer Magnetic Stirrer
Microstirrer Magnetic Stirrer
Microstirrer Magnetic Stirrer
Fisherbrand
Microstirrer Magnetic Stirrer
Small magnetic stirrer specially designed for microtitrations.
Have Questions?
Catalog NumberCapacity
FKS117656945 L
Catalog number FKS11765694
Price (MYR)
523.00
EA
Add to cart
Capacity:
5 L
Price (MYR)
523.00
EA
Add to cart
Ensures a high resistance to chemical agents. Fisherbrand™ Microstirrer Magnetic Stirrer is small, simple and efficient while remaining cold even after several days of continuous use.
High Reliability:
  • Control panel is shielded from liquid spills
  • Epoxy-coated tough steel casing
  • Long life operation
  • Accepts up to 5 liter flasks
Applications in Mind:
  • Pale surface for visualization of particles and color changes
  • Remains cold, even after days of continuous use
  • Specifically designed for microtitration
  • Excellent speed control, even at low speed
Simple and Practical:
  • Speed regulation up to 1100 rpm
  • Universal mains voltage / frequency range
  • Light weight
Specifications
Speed1100 rpm
Stirring Positions1
Top Plate MaterialEpoxy
Capacity5 L
DisplayAnalog
ShapeSquare
Voltage100/240 V
Wattage0.6 w
Dimensions (L x W x H)48 x 120 x 128 mm (H x W x D)
Height (Metric)48 mm
Width (Metric)120 mm
Electrical Requirements100 to 240 V, 50/60 Hz, 0.6 w
Frequency50/60 Hz
MaterialMetal
Speed ControlExcellent speed control even at low revolutions
TypeMagnetic Stirrer
Weight (Metric)0.55 kg
Unit SizeEA