Stainless Steel Spoon One End PTFE End Spatulas
Actual product may vary
Fisherbrand
Stainless Steel Spoon One End PTFE End Spatulas
Get superior results when you use the combination spoon and spatula for your essential laboratory work. Made of stainless steel and PTFE-coated, the Fisherbrand™ Stainless Steel Spoon One End Spatula PTFE End is ideal for handling chemicals and prevents loss of critical quantities due to sticking.
Have Questions?
Catalog NumberLength (Metric)
FKS11583512150 mm
Catalog number FKS11583512
Price (MYR)
-
Length (Metric):
150 mm
  • Double-ended spatula
  • Single-coated PTFE that prevents build up on surfaces 
  • Prevents loss of critical quantities due to sticking
  • Fully covered PTFE coating—ideal for handling chemicals
  • 304 Stainless Steel PTFE Coated 
  • Length 120mm long
  • Spoon measurement 20mm wide
  • Spoon capacity (liquid) = 2ml
Specifications
Quantity1 Ea.
Capacity (Metric)2 mL
CoatingPTFE Xylan™
Product TypeSpatula
For Use With (Application)Ideal for Biology, Microbiology and Pharmaceutical Applications
Width (Metric)20 mm
Dimensions (L x W)150 x 20 mm
Temperature (Metric) Max-195°C to +260°C
MaterialStainless Steel
Length (Metric)150 mm
Unit SizeEach