Polygon Spinbar™ Magnetic Stirring Bars without Pivot Ring
Polygon Spinbar™ Magnetic Stirring Bars without Pivot Ring
Actual product may vary
Bel-Art
Polygon Spinbar™ Magnetic Stirring Bars without Pivot Ring
Adds turbulence relative to similar smooth size cylindrical bars. Manufactured in a registered ISO 9001:2008 facility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Length (English)Diameter (English)
FIS22-33144750.8 mm9.5 mm2 in.0.37 in.
FIS22-33141315.9 mm7.9 mm0.62 in.0.31 in.
FIS22-33143938 mm9.5 mm1.5 in.0.37 in.
Catalog number FIS22-331447
Price (MYR)
1,736.00
EA
Add to cart
Length (Metric):
50.8 mm
Diameter (Metric):
9.5 mm
Length (English):
2 in.
Diameter (English):
0.37 in.
Price (MYR)
1,736.00
EA
Add to cart
Adds turbulence relative to similar smooth size cylindrical bars. Manufactured in a registered ISO 9001:2008 facility.
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low-friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility
  • Autoclave safe
Specifications
AutoclavableAutoclavable
Certifications/ComplianceFDA, USP Class VI, ISO 9001:2008
ColorWhite
Diameter (English)0.37 in.
Length (English)2 in.
CoatingPTFE, FDA-grade, USP Class VI
Diameter (Metric)9.5 mm
Length (Metric)50.8 mm
MaterialAlnico Magnet
Quantity1
ShapePolygon
TypeMagnetic Stir Bar
Unit SizeEA