▸
Personal Protective Equipment
▸
Eye Protection and Face Protection
▸
General Purpose Safety Glasses
▸
North™ Norton 180™ T1800 Series Safety Glasses
Actual product may vary
Honeywell
North™ Norton 180™ T1800 Series Safety Glasses
Molded polycarbonate wraparound safety glasses
Have Questions?
Catalog Number
Lens Tint
FIS17-230A
Clear
Catalog number
FIS17-230A
Price (MYR)
-
Lens Tint:
Clear
Scratch-resistant lens absorbs 99.9% of UV radiation up to 380nm
One-piece, impact-resistant lens offers a large field of vision
Frosted browguard reduces glare from overhead lighting
Vented sideshields
Specifications
Lens Coating
Hardcoat
Lens Tint
Clear
Product Type
Safety glasses
For Use With (Equipment)
Norton 180 T1800 Series Safety Glasses
Lens Material
Molded polycarbonate