TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology, Fisher BioReagents™
TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology, Fisher BioReagents™
Fisher Chemical
TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology, Fisher BioReagents™
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Packaging:
View all
Nalgene Poly Bottle
Catalog number FGCBP2471-1
Price (MYR)
0.00
EA
Add to cart
Quantity:
1 L
Packaging:
Nalgene Poly Bottle
Request bulk or custom format
Price (MYR)
0.00
EA
Add to cart
Chemical Identifiers
CAS7447-40-7, 7647-01-0, 7647-14-5, 77-86-1, 7732-18-5
Specifications
Chemical Name or MaterialTris Buffered Saline
Name Note10X Solution
CAS7447-40-7
ChemAlert Storage SymbolGray
Boiling Point100°C
View more
TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology
Washing buffer for alkaline phosphatase or peroxidase conjugates in Western blotting