▸
Salts and Inorganics
▸
Inorganic Salts
▸
Transition Metal Salts
▸
Ferrous Sulfate Heptahydrate (Crystalline), ACS Grade, Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Ferrous Sulfate Heptahydrate (Crystalline), ACS Grade, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMI146-500
500 g
Poly Bottle
Catalog number
FCMI146-500
Price (MYR)
518.00
Each
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
518.00
Each
Add to cart
Specifications
Boiling Point
300°C
Chemical Name or Material
Ferrous Sulfate Heptahydrate
Melting Point
64°C
CAS
7782-63-0
ChemAlert Storage Symbol
Gray
View more