DMF, Dimethylformamide, GC Headspace Grade, Thermo Scientific™
DMF, Dimethylformamide, GC Headspace Grade, Thermo Scientific™
DMF, Dimethylformamide, GC Headspace Grade, Thermo Scientific™
DMF, Dimethylformamide, GC Headspace Grade, Thermo Scientific™
Fisher Chemical
DMF, Dimethylformamide, GC Headspace Grade, Thermo Scientific™
DMF processed to high purity for accurate and repeatable determination of trace levels of residual solvents.
Have Questions?
Catalog NumberQuantityPackaging
FCMD133-11 LGlass Bottle
Catalog number FCMD133-1
Price (MYR)
968.00
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
968.00
Each
Specifications
Chemical Name or MaterialDMF, Dimethylformamide
Absorbance0.300 AU Maximum (275nm) , 0.050 AU Maximum (300nm), 0.005 AU Maximum (350nm), 0.005 AU Maximum (400nm)
CAS68-12-2
ColorColorless
Recommended StorageRoom temperature, packaged under inert atmosphere.
View more
  • Every lot is tested to ensure interference free baselines for interference free determination of Class 1, 2, and 3 residual solvents
  • 1 ppm max. of Class 1 Solvents, 10 ppm max. of Class 2 solvents, and 50 ppm max of Class 3 residual solvents
  • This product is UV Absorbance Tested to ensure low organic contamination
  • Processed for low water content to facilitate the extraction of organic volatile impurities, or VOC