Agarose (Protein Electrophoresis Grade), Fisher BioReagents™
Agarose (Protein Electrophoresis Grade), Fisher BioReagents™
Agarose (Protein Electrophoresis Grade), Fisher BioReagents™
Fisher Chemical
Agarose (Protein Electrophoresis Grade), Fisher BioReagents™
Have Questions?
Catalog NumberQuantityPackaging
FBRBP164-500500 gPoly Bottle
Catalog number FBRBP164-500
Price (MYR)
3,752.00
Each
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
3,752.00
Each
Specifications
Chemical Name or MaterialAgarose
Physical FormPowder/Solid
CAS9012-36-6
ChemAlert Storage SymbolGray
Recommended StorageRT
View more
Agarose (Protein Electrophoresis Grade)