▸
Gel Electrophoresis Equipment and Supplies
▸
Gel Electrophoresis Reagents and Buffers
▸
Electrophoresis Gel Casting Reagents
▸
Agarose (Protein Electrophoresis Grade), Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Agarose (Protein Electrophoresis Grade), Fisher BioReagents™
CAS:
9012-36-6 |
Molecular Formula:
C
12
H
18
O
9
Have Questions?
Change view
Quantity:
25 g
100 g
500 g
Packaging:
Poly Bottle
Catalog number
FBRBP164-25
Price (MYR)
609.00
Each
Add to cart
Quantity:
25 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
609.00
Each
Add to cart
Specifications
Chemical Name or Material
Agarose
Physical Form
Powder/Solid
CAS
9012-36-6
ChemAlert Storage Symbol
Gray
Recommended Storage
RT
View more
Agarose (Protein Electrophoresis Grade)