Ethylenediaminetetraacetic Acid, Di Na Salt Dihydr. (Crystalline Powd./Electrophor.), Fisher BioReagents™
Ethylenediaminetetraacetic Acid, Di Na Salt Dihydr. (Crystalline Powd./Electrophor.), Fisher BioReagents™
Ethylenediaminetetraacetic Acid, Di Na Salt Dihydr. (Crystalline Powd./Electrophor.), Fisher BioReagents™
Fisher Chemical
Ethylenediaminetetraacetic Acid, Di Na Salt Dihydr. (Crystalline Powd./Electrophor.), Fisher BioReagents™
For minimization of metal ion impurities in reaction buffers
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
View all
Glass Bottle
Catalog number FBRBP120-500
Price (MYR)
619.00
Each
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
619.00
Each
Specifications
Chemical Name or MaterialEthylenediamine Tetraacetic Acid, Disodium Salt Dihydrate
CAS6381-92-6
ChemAlert Storage SymbolGray
Health Hazard 2CAUTION!
Decomposition Information>252°C
View more
For minimization of metal ion impurities in reaction buffers
The disodium salt of EDTA is a chelating agent for metal ions and is used extensively in molecular biology to minimize metal ion impurities in reaction buffers.