▸
Facility Safety and Maintenance
▸
Cleaning Supplies and Equipment
▸
Wipes and Wipe Accessories
▸
Controlled Environments Wipes
▸
EasyReach™ Cleaning Pads
Actual product may vary
Contec
EasyReach™ Cleaning Pads
EasyReach™ Cleaning Pads are very absorbent, which ensures even coverage of disinfectant or IPA.
Have Questions?
Change view
Catalog Number
Dimensions (L x W)
Product Type
Material
Sterility
CTCAMEC0113
7.5 x 2.8 in. (19 x 7 cm)
Cleaning Pad
Quilted Polyester
Sterile
CTCMEQT0002SZ
7.5 x 2.8 in. (19 x 7 cm)
Cleaning Pad
Quilted Polyester
Sterile
2 Options
Catalog number
CTCAMEC0113
Price (MYR)
-
Dimensions (L x W):
7.5 x 2.8 in. (19 x 7 cm)
Product Type:
Cleaning Pad
Material:
Quilted Polyester
Sterility:
Sterile
Single-use sealed polyester
Provide a low level of particulates which is essential for use in product contact areas
Very absorbent, ensuring good even coverage of disinfectant or IPA
Easy grab edge on the cleaning pad ensures the pad fits tightly on the mop frame
Sterile cleaning pads with dry and presaturated options
Specifications
Product Type
Cleaning Pad
Length (English)
7.5 in.
Length (Metric)
19 cm
Width (English)
2.8 in.
Width (Metric)
7 cm
Material
Quilted Polyester
Autoclavable
Not Autoclavable
Color
White
Disposable
Single-use
Dimensions (L x W)
7.5 x 2.8 in. (19 x 7 cm)
For Use With (Equipment)
Cleaning Isolators, LAF Hoods, Glove Boxes and Other Mini Environments
Packaging Quantity
4/Bag, 4 Pads/Bag, 15 Bags/Cs.
Sterility
Sterile