▸
Personal Protective Equipment
▸
Respiratory Protection
▸
Air-Purifying Respirators
▸
Powered Air Purifying Respirators
▸
PAPR Replacement Parts and Accessories
▸
Unit ID Panels
Actual product may vary
CleanSpace
Unit ID Panels
Identification panels are available in assorted colors: blue, green, rose, silver. Supplied in packs of 6.
Have Questions?
Catalog Number
Quantity
CSECS3013
6/Pk.
Catalog number
CSECS3013
Price (MYR)
387.45
Pack
Add to cart
Quantity:
6/Pk.
Price (MYR)
387.45
Pack
Add to cart
Assorted colors: blue, green, rose, silver.
Supplied in packs of 6.
Specifications
Disposable
Reusable
Product Type
ID Badges
For Use With (Equipment)
CS3000 PAPR System
Model
CS3013
Quantity
6/Pk.