Forma&trade; Steri-Cult&trade; CO<sub>2</sub> Incubator, 232L &amp; 323L
Actual product may vary
Corning
Forma™ Steri-Cult™ CO2 Incubator, 232L & 323L
Carry out high-volume culturing with a high-level of contamination control with these stackable CO2 incubators, which feature microprocessor controls.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVoltageCapacity (Metric)
CRG3311230 V322.8 L
3311
also known as BP3311
230 V322.8 L
Catalog number CRG3311
Price (MYR)
6,104.00
Case
Voltage:
230 V
Capacity (Metric):
322.8 L
Price (MYR)
6,104.00
Case
Carry out high-volume culturing with a high-level of contamination control with these stackable Thermo Scientific™ Forma™ Steri-Cult™ CO2 Incubators. This incubator combines the most advanced components available into a single incubator: CO2 sensor, precise microprocessor controls, active humidity control, HEPA air filtration and high-temperature decontamination capability.
Direct Heat Chamber
  • The high capacity chamber are designed to provide maximum culturing space for high throughput applications
  • Standard right hand door swing
  • Polished stainless steel interior with coved corners is designed to be easy-to-clean
  • Active airflow gently distributes air throughout the chamber ensuring homogeneous conditions
  • Gentle airflow circulation is designed to reduce the risk of cross contamination
  • Sturdy stainless steel shelves and supports can be removed without tools for easy cleaning or adjustment
  • IR (Infrared) CO2 sensor provides precise monitoring and is unaffected by changes in rH setpoint 
Active Humidity Control
  • Humidity level is easily adjustable with no water pan to manage
  • External water reservoir is designed to minimize chamber contamination and allows refilling without opening chamber
  • At-a-glance water level indicator helps prevent sample desiccation 
140°C Sterilization Cycle
  • On-demand 140°C overnight (14 hours) sterilization cycle is independently validated
  • Simplifies cleaning and eliminates the need for separate autoclaving and reassembly of components
  • Designed to effectively eliminate bacteria, mold, yeast, and mycoplasmas from internal surfaces 
ISO Class 5 HEPA Filtration Airflow System
  • In-chamber HEPA air filtration continuously filters entire chamber volume every 60 seconds
  • Reaches ISO Class 5 air quality in 5 minutes after door opening
  • Continuous protection minimizes contaminants settling on interior surfaces
Enviro-Scan Microprocessor Control
  • Easy-to-use, alphanumeric LED display
  • Flexible, intuitive controls with message center including process and alarm status messages
  • Configurable audible on/off, access code, tracking low and high alarms
Specifications
Capacity (English)11.4 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data OutputsOptional 4-20mA
DescriptionSingle 323L incubator
Capacity (Metric)322.8 L
Chamber MaterialPolished Stainless Steel
CO2 Concentration Range0 to 20%
CO2 Sensor TechnologyIR Sensor
Convection TypeWater-jacketed
LightingLED
MaterialStainless Steel
No. of Places22
No. of Shelves5 std./22 max.
Oxygen ControlNone
Relative Humidity95%, Controllable
Temperature Range (Metric)Ambient +5° to 50°C
TypeCO2 Incubator
Humidity SourceExternal humidity reservoir
Dimensions (D x W x H) Interior20.6 x 28.8 x 32.8 in. (52.3 x 73.2 x 83.8 cm)
Dimensions (L x W x H)27 x 43 x 39.4 in. (68.6 x 109.2 x 100.2 cm)
Weight (English)410 lb.
Electrical Requirements230 V, 50/60 Hz