Coliform Sample Container
Actual product may vary
Corning
Coliform Sample Container
Automatically dechlorinates sample
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
CRG1700-100Sodium Thiosulfate Tablet
CRG1705-100w/o Sodium Thiosulfate Tablet
FIS09-730-91Aw/o Sodium Thiosulfate Tablet
Catalog number CRG1700-100
Price (MYR)
552.00
Case
Includes:
Sodium Thiosulfate Tablet
Price (MYR)
552.00
Case
These Corning™ containers are used to test nonchlorinated drinking water.
  • Manufactured from pure, recyclable polypropylene.
  • Capacity: 120 mL total capacity.
  • Capacity at line: 100 mL ± 2.5 mL (2.5%).
  • The one-piece container has attached lid to reduce chance of contamination.
  • Locking arrow assures sterility has not been compromised.
  • Leak tight when sealed properly. An added benefit is the tie-down which protects against accidental opening.
  • Sterile-by-process
Specifications
Certifications/ComplianceEPA
Closure ColorNatural
Closure MaterialPolypropylene
Closure TypeSnap Cap
Volume (Metric)120 mL
DisposableSingle-use
For Use With (Application)Test for the presence of coliform bacteria, microbiologic contaminants in drinking water
GraduatedYes
IncludesSodium Thiosulfate Tablet
Neck StyleWide Mouth
SterilitySterile
Volume (English)4 oz.
Height (English)3.77 in.
Height (Metric)9.6 cm
Diameter (English) Outer1.82 in.
Diameter (Metric) Outer4.64 cm
Unit SizeCase