1-16K Refrigerated Laboratory Microcentrifuge
Actual product may vary
Corning
1-16K Refrigerated Laboratory Microcentrifuge
Makes lab work easier. Sigma Laborzentrifugen 1-16K Refrigerated Laboratory Microcentrifuge has ergonomic design.
Have Questions?
Catalog NumberIncludes
CRG10030PP rotor
Catalog number CRG10030
Price (MYR)
3,987.00
Case
Includes:
PP rotor
Price (MYR)
3,987.00
Case
  • Capacity: 24 x 1.5/2.2mL
  • Precooling of rotors during standstill, refrigerant CFC-free (R 134a)
  • Max speed 15,000rpm (20.627g)
  • Guaranteed 4°C at max speed
  • 2 acceleration/deceleration curves
  • Last run memory
  • Stainless steel bowl
  • Emergency lid lock release
  • Motorised lid lock, auto lid open
  • Maintenance free brushless drive motor
  • Over-speed controller
  • Window in lid for external speed control
  • Manufactured according to IEC 61010, RoHS compliant
Specifications
Capacity24 x 1.5/2.2 mL
Max. Speed15,000 rpm
Noise Level58 dB
Certifications/ComplianceIEC 61010, RoHS compliant
IncludesPP rotor
Electrical Requirements220/240 V 50/60 Hz
Depth (English)25.9 in.
Depth (Metric)660 mm
Wattage460 w
RefrigeratedYes
Profile (Acceleration/Braking)15/13 sec.
Run Time10 sec. to 99 min. 59 sec.
Program Storage10 programs
Product Line1-16K Refrigerated Microcentrifuge
Product Type1-16K Refrigerated Microcentrifuge
Temperature Range (Metric)4°C
Unit SizeCase