Poly(4-vinylpyridine), 25% crosslinked with divinylbenzene, ca. 18 mesh
Poly(4-vinylpyridine), 25% crosslinked with divinylbenzene, ca. 18 mesh
Poly(4-vinylpyridine), 25% crosslinked with divinylbenzene, ca. 18 mesh
Poly(4-vinylpyridine), 25% crosslinked with divinylbenzene, ca. 18 mesh
Acros Organics
Poly(4-vinylpyridine), 25% crosslinked with divinylbenzene, ca. 18 mesh
CAS: 9017-40-7 | (C7H7N)n | 235.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Catalog number ACR460670500
Price (MYR)
-
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPoly(4-vinylpyridine)
CAS9017-40-7
Flash Point>150°C
Infrared SpectrumAuthentic
Molecular Formula(C7H7N)n