▸
Organic compounds
▸
Organooxygen compounds
▸
Carbonyl compounds
▸
Complex Aldehydes
▸
3-(Trifluoromethoxy)benzaldehyde, 95%, Thermo Scientific™
Element Search
Structure Search
Acros Organics
3-(Trifluoromethoxy)benzaldehyde, 95%, Thermo Scientific™
Have Questions?
Change view
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number
ACR297650050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
3-(Trifluoromethoxy)benzaldehyde
Name Note
95%
CAS
52771-21-8
CAS Min %
94.0
Health Hazard 1
GHS Signal Word:
Warning