Triton™ X-100, Thermo Scientific™
Triton™ X-100, Thermo Scientific™
Triton™ X-100, Thermo Scientific™
Acros Organics
Triton™ X-100, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
2.5 L
10 L
Packaging:
View all
EcoSafPak Glass Bottle
Catalog number ACR215680010
Price (MYR)
-
Quantity:
1 L
Packaging:
EcoSafPak Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTriton™ X-100
CAS9002-93-1
Health Hazard 1Danger
Boiling Point270.0°C
Health Hazard 2GHS H Statement
Harmful to aquatic life with long lasting effects.