Evolution™ Peltier Thermostatting Accessories
Actual product may vary
Evolution™ Peltier Thermostatting Accessories
Evolution™ Peltier Thermostatting Accessories
Thermo Scientific
Evolution™ Peltier Thermostatting Accessories
Achieve superior performance in temperature control and sample stirring for many UV-Visible analysis methods.
Have Questions?
Catalog NumberFor Use With (Equipment)TypeIncludes
699-131200Evolution spectrophotometers8-Cell Peltier SystemPeltier Cooling Unit, 8-Sample
Catalog number 699-131200
HSN 90279090
Price (MYR)
82,376.00
Each
For Use With (Equipment):
Evolution spectrophotometers
Type:
8-Cell Peltier System
Includes:
Peltier Cooling Unit, 8-Sample
Price (MYR)
82,376.00
Each
Thermo Scientific™ Peltier Accessories for the Thermo Scientific™ Evolution™ One, Evolution™ One Plus and Evolution™ Pro UV-Vis Spectrophotometers provide thermostatting and ramping with precise temperature control which is important for many UV-Visible analysis methods. Outperforming traditional heater/chiller units, these accessories offer thermostatting and ramping with precise temperature control between 0 °C and 110 °C. Add superior performance with compact integrated design, simple set-up, and fast, precise temperature control.
Precise Thermostatting
Peltier Accessories for the Evolution UV-Visible Spectrophotometers utilize re-circulating water as an effective heat sink to allow the Peltier element to heat or cool the block over a wide temperature range. The single-cell holder can reach as low as 0 °C and as high at 110 °C. Thermo Scientific offers 3 Peltier options to cover many possible applications, including a multi-cell unit. Stirring control for the single cell holders is provided when you place a micro-stir bar inside the cuvette. Unlike thermostatting with a traditional re-circulating water bath where a large volume of liquid must be heated or cooled, heating or cooling the cell holder with the Peltier effect means more rapid and more precise temperature control. Choose between water or chemical coolant to best fit your needs.
Temperature Ramping
Thermo Scientific INSIGHT Pro Software allows you to control and monitor the block temperature of the Peltier unit and allows you to implement programmed temperature ramping with multiple ramp and hold cycles and variable temperature kinetics.  Add a temperature probe hub and probes, and you can monitor the exact temperature in each cell as you perform heating and cooling cycles. The Insight Pro software also includes pre-set methods that can be used to study DNA denaturation and renaturation or other complex temperature programmed experiments.
Interested in instrument and Peltier bundles? Click here for more information.
Specifications
Description8 Cell Peltier System for Evolution Spectrophotometers
Type8-Cell Peltier System
For Use With (Equipment)Evolution spectrophotometers
IncludesPeltier Cooling Unit, 8-Sample
Measurement RangeRamp rate: 0.4-20 °C
Temperature Precision0.1°C
Temperature Range (Metric)5°C to 100°C
Unit SizeEach