Chest Freezer Racks
Chest Freezer Racks
Actual product may vary
Chest Freezer Racks
Chest Freezer Racks
Chest Freezer Racks
Thermo Scientific
Chest Freezer Racks
Use Thermo Scientific™ Chest Freezer Racks with Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsFor Use With (Equipment)
398186Boxes (2 in.)13, 17, 20 cu. ft. Freezers
189001General Freezer Items12.7 cu. ft. Freezers
398184Boxes (2 in.)3 cu. ft. Chest Freezers
Catalog number 398186
HSN 73269099
Price (MYR)
1,115.00
EA
Add to cart
Holds:
Boxes (2 in.)
For Use With (Equipment):
13, 17, 20 cu. ft. Freezers
Price (MYR)
1,115.00
EA
Add to cart
Use Thermo Scientific™ Chest Freezer Racks with TDE Series Ultra-Low Temperature Chest Freezers as well as legacy Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
  • Warranty varies by country, please contact us for details
Specifications
ColorSilver
HoldsBoxes (2 in.)
MaterialStainless Steel
Positions12 Boxes/Rack
For Use With (Equipment)13, 17, 20 cu. ft. Freezers
Freezer Compatibility (Capacity)13, 17, 20 cu. ft.
Width (English)5.5 in.
Height (English)26.5 in.
Depth (English)5.5 in.
Depth (Metric)14 cm
Product TypeSide Access Rack Tower
Height (Metric)67.3 cm
Width (Metric)14 cm
Unit SizeEA