Propyl propionate, 98+%
Propyl propionate, 98+%
Propyl propionate, 98+%
Acros Organics
Propyl propionate, 98+%
CAS: 106-36-5 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Glass bottle
Catalog number 385520010
HSN 29155000
Price (MYR)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS106-36-5
IUPAC Namepropyl propanoate
Molecular FormulaC6H12O2
InChI KeyMCSINKKTEDDPNK-UHFFFAOYSA-N
SMILESCCCOC(=O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=98.0 %
Refractive index1.3820 to 1.3940 (20°C, 589 nm)
Appearance (Form)Liquid