Filters and Accessories for Thermo Scientific Multiskan™ FC Microplate Readers
Filters and Accessories for Thermo Scientific Multiskan™ FC Microplate Readers
Actual product may vary
Filters and Accessories for Thermo Scientific Multiskan™ FC Microplate Readers
Filters and Accessories for Thermo Scientific Multiskan™ FC Microplate Readers
Thermo Scientific
Filters and Accessories for Thermo Scientific Multiskan™ FC Microplate Readers
Multiskan™ FC microplate photometers can be equipped with wide range of additional filters to expand the instrument functionality and application flexibility.
Have Questions?
Catalog NumberProduct TypeWavelength
1424505Optical Filter450 nm
Catalog number 1424505
HSN 90029000
Price (MYR)
1,718.00
Each
Add to cart
Product Type:
Optical Filter
Wavelength:
450 nm
Price (MYR)
1,718.00
Each
Add to cart
Multiskan FC brings together over 40 years' experience of the well-known Multiskan product brand, with features for enhanced usability. The Multiskan FC has a 340–850 nm wavelength range, enabling a wide variety of applications from enzyme kinetic studies to Lowry assays. Extensive choice of filters for optimal wavelength selection for assays increases the application flexibility.
  • Multiskan FC is equipped with an eight-position filter wheel with three preinstalled standard filters, 405 nm, 450 nm and 620 m.
  • A comprehensive range of easy-to install additional filters is available for order
  • Superior usability and logical workflow with Thermo Scientific™ SkanIt™ Software
Specifications
Product TypeOptical Filter
For Use With (Equipment)Multiskan FC
Product LineMultiskan
Wavelength450 nm
Unit SizeEach