▸
Pumps
▸
Vacuum Pumps
▸
CRVpro™ Starter Package
Actual product may vary
Welch
CRVpro™ Starter Package
The robust vacuum pump CRVpro belongs to the new high-performance two-stage rotary vane pumps generation of Welch.
Have Questions?
Catalog Number
Model
WEL330073
CRVpro 8
Catalog number
WEL330073
Price (MYR)
-
Model:
CRVpro 8
CRVpro Starter Package Includes:
Oil mist separator AKD 16
2-way ball valve
Hose connector DN 16KF-10/8 mm
Vacuum hose (1.5 m)
Oil change set
Specifications
Includes
CRVpro with Oil Mist Separator AKD 16, 2-Way Ball Valve, Hose Connector, Vacuum Hose (1.5 m), Oil Change Set
Model
CRVpro 8
Pump Type
Rotary Vane Vacuum Pump