Electronic Ice Bucket Ceramic Beads
Product Image
Fisherbrand
Electronic Ice Bucket Ceramic Beads
Keep your samples cold, clean, and dry
Have Questions?
Catalog number FKS11810615
Price (MYR)
382.00
EA
Add to cart
Price (MYR)
382.00
EA
Add to cart
These Techne™ N°ICE ceramic-coated beads maintain the temperature and support the sample vessels for as long as necessary. The beads are chemically resistant and if required can be autoclaved at 134°C. As a time saving option, the bucket of beads can be incubated in the fridge overnight and simply inserted into the unit as and when required. Compatible with the 24120-00 Electronic Ice Bucket, sold separately.
Specifications
TypeReplacement Ceramic Beads
Unit SizeEA