▸
Microscope Slides
▸
Glass Microscope Slides
▸
Microscope Slides with Ground Edges
Actual product may vary
Fisherbrand
Microscope Slides with Ground Edges
Wrapped in cellophane for improved cleanliness. These Microscope Slides with Ground Edges are washed and polished glass slides with 90° ground edge.
Have Questions?
Change view
Catalog Number
Thickness (Metric)
Frosted
FKS11562203
0.8 to 1 mm
Twin Frosted
FKS12373118
0.8 to 1 mm
Not Frosted
FKS12383118
1 to 1.2 mm
Not Frosted
FKS11572203
1 to 1.2 mm
Twin Frosted
4 Options
Catalog number
FKS11562203
Price (MYR)
35.00
Pack
Add to cart
Thickness (Metric):
0.8 to 1 mm
Frosted:
Twin Frosted
Price (MYR)
35.00
Pack
Add to cart
Glass 76 mm x 26 mm
Available in fully clear glass or frosted at one end
Ground on all four edges
Specifications
Edges
90°, Ground
Frosted
Twin Frosted
Length (Metric)
76 mm
Material
Glass
Precleaned
Yes
Thickness (Metric)
0.8 to 1 mm
Type
Plain Microscope Slide
Width (Metric)
26 mm
Color
Clear
Quantity
50 Pk.