Check Order Status
IPA (Isopropyl Alcohol), 70% v/v, Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
IPA (Isopropyl Alcohol), 70% v/v, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMA459-1
1 L
Poly Bottle
Catalog number
FCMA459-1
Price (MYR)
560.00
Each
Add to cart
Quantity:
1 L
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
560.00
Each
Add to cart
Specifications
Chemical Name or Material
IPA (Isopropyl Alcohol), 70% v/v
Melting Point
-88°C
CAS
67-63-0
Color
Colorless
Health Hazard 2
WARNING!
View more