▸
Chromatography Supplies
▸
Chromatography Fittings
▸
FastPack Inlet Kits for Agilent GCs
Actual product may vary
Masterflex
FastPack Inlet Kits for Agilent GCs
Includes all parts needed for routine maintenance of injection ports and the entire system.
Have Questions?
Catalog Number
Type
EPP6734000012
Splitless Inlet Maintenance Kit
Catalog number
EPP6734000012
Price (MYR)
-
Type:
Splitless Inlet Maintenance Kit
Economical - costs less than the sum of the individual parts
Factory sealed Mylar bag prevents contamination of products from weeks in the lab
Each bag includes five maintenance kits
Includes: inlet liner (choose from three popular styles), Viton O-ring, 0.8mm ID gold-plated inlet seal and washer, 11mm Thermolite septum
Specifications
Type
Splitless Inlet Maintenance Kit
Includes
washer, septa and liner
For Use With
Agilent GC