P/S™ Precision Pump Tubing, PharMed™ BPT
P/S™ Precision Pump Tubing, PharMed™ BPT
Actual product may vary
Eppendorf
P/S™ Precision Pump Tubing, PharMed™ BPT
Use this tubing for cell and tissue culture work and pharmaceutical applications. Masterflex™ L/S™ High-Performance Tubing, PharMed™ BPT resists acids, alkalies, oxidizing agents, and animal and vegetable oils. Tubing is heat-sealable, bondable and formable. Good for light-sensitive samples.
Have Questions?
Catalog NumberDiameter (Metric) InnerFlow Rate
EPP67310000124.8 mm1.8 to 1100mL/min.
Catalog number EPP6731000012
Price (MYR)
-
Diameter (Metric) Inner:
4.8 mm
Flow Rate:
1.8 to 1100mL/min.
Masterflex™ L/S™ pump tubing is manufactured to extremely close tolerances that match our L/S™ pump heads, ensuring accurate, repeatable flow and long tubing life. Our pump tubing is factory-tested and optically inspected to provide the best performance from your peristaltic pump.
  • Applications: Cell and tissue culture work and pharmaceutical uses. Also good for light-sensitive samples.
  • Characteristics: Nontoxic; nonhemolytic; extremely low permeability; resists acids, alkalies, oxidizing agents, and animal and vegetable oils; heat-sealable, bondable, and formable. Opaque beige.
  • Certifications: FDA compliant and meets USP Class VI, EP, and NSF-51 requirements. Tubing is manufactured according to GMP.
Specifications
Certifications/ComplianceFDA 21 CFR 175.300
AutoclavableAutoclavable
Diameter (English) Inner19/100 in.
Diameter (Metric) Inner4.8 mm
MaterialPharMed
Diameter (English) Outer0.59 in.
Diameter (Metric) Outer15 mm
Overall Length (English)25 ft.
Overall Length (Metric)1520 cm
For Use WithFH15 and FH30 Peristaltic pump
Min. Operating Temperature (English)-60°F
Min. Operating Temperature (Metric)-51°C
Resistance PropertiesAcids, Alkalies
SterilitySterile
Tubing Size15
Flow Rate1.8 to 1100mL/min.
Hose Barb0.18 in.
Height (Metric)8.8 m Water
Height (English)29 ft. Water
Gas PermeabilityCO2: 360, H2: 97, O2: 80, N2: 40
ColorOpaque Beige
Max. Operating Temperature (English)270°F
Max. Operating Temperature (Metric)132°C
Product TypePump Tubing
Unit SizeEA