CONSTIX™ Cotton Swabs
CONSTIX™ Cotton Swabs
Actual product may vary
Corning
CONSTIX™ Cotton Swabs
Manufactured with the longest staple cotton fibers and a simple methyl-cellulose binder. CONSTIX™ Cotton Swabs provide very low levels of extractable residue, particles and fibers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) TipLength (Metric) HandleLength (Metric) TipShape
CTCSC-15 mm76.2 mm13 mmPointed double ended
CTCPNHS-99DX5 mm76.2 mm13 mmPointed double ended
Catalog number CTCSC-1
Price (MYR)
-
Diameter (Metric) Tip:
5 mm
Length (Metric) Handle:
76.2 mm
Length (Metric) Tip:
13 mm
Shape:
Pointed double ended
  • Tightest, smoothest and cleanest cotton swabs available
  • Absorbent to most common solvents including isopropyl alcohol and acetone
  • Use in critical applications including disk drive manufacturing and high-level optical
  • Ideal for use with aqueous-based solutions
Specifications
Diameter (English) Handle0.1 in.
Length (English)3.51 in.
For Use With (Application)For use in some of the most critical applications, including disk drive manufacturing and high-level optical surfaces
Product TypeSwab
Diameter (Metric) Handle2.5 mm
Length (English) Handle3 in.
Length (Metric) Handle76.2 mm
MaterialCotton
ShapePointed double ended
Length (English) Tip0.51 in.
Length (Metric) Tip13 mm
Width (Metric) Tip8 mm
Width (English) Tip0.5 in.
Diameter (Metric) Tip5 mm
Diameter (English) Tip0.2 in.
SterilityNon-sterile
Adhesive TypeAqueous-based
Handle MaterialWound Paper
Tip StylePointed
Tip MaterialCotton
Unit SizeCS