Polynit™ Wipes
Polynit™ Wipes
Actual product may vary
Polynit™ Wipes
Polynit™ Wipes
Corning
Polynit™ Wipes
Clean surfaces of products and process tools. The Polynit™ Wipes are a durable option for ISO Class 5 environments.
Have Questions?
Catalog NumberDimensions (L x W)Packaging QuantitySterility
CTCPN-44B10 x 10 cm (4 x 4 in.)60 Wipes/Bag (Bulk)Non-sterile
Catalog number CTCPN-44B
Price (MYR)
-
Dimensions (L x W):
10 x 10 cm (4 x 4 in.)
Packaging Quantity:
60 Wipes/Bag (Bulk)
Sterility:
Non-sterile
  • For general-purpose cleaning in ISO Class 5 (FED-STD-209E Class 100/M3.5) environments
  • Ideal for wiping exterior surfaces of products and cleaning interior surfaces of process tools or other equipment
Specifications
Product TypeWipe
Length (English)4 in.
Length (Metric)10 cm
Width (English)4 in.
Width (Metric)10.16 cm
MaterialPolyester
AutoclavableNot Autoclavable
Certifications/ComplianceISO Class 5
ColorWhite
DisposableSingle-use
Dimensions (L x W)10 x 10 cm (4 x 4 in.)
For Use With (Application)Ideal for general purpose cleaning and wiping exterior surfaces of products
Packaging Quantity60 Wipes/Bag (Bulk)
Ply2-ply