Sibutramine hydrochloride monohydrate
Sibutramine hydrochloride monohydrate
Sibutramine hydrochloride monohydrate
Sibutramine hydrochloride monohydrate
Acros Organics
Sibutramine hydrochloride monohydrate
CAS: 125494-59-9 | C17H29Cl2NO | 334.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ACR462200050
Price (MYR)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSibutramine hydrochloride monohydrate
CAS125494-59-9
Molecular FormulaC17H29Cl2NO
Quantity5 g
InChI KeyKFNNPQDSPLWLCX-UHFFFAOYNA-N