Dowex™ 50WX8, 200-400 mesh, ion-exchange resin
Dowex™ 50WX8, 200-400 mesh, ion-exchange resin
Dowex™ 50WX8, 200-400 mesh, ion-exchange resin
Dowex™ 50WX8, 200-400 mesh, ion-exchange resin
Acros Organics
Dowex™ 50WX8, 200-400 mesh, ion-exchange resin
CAS: 11119-67-8
Have Questions?
Catalog NumberQuantityPackaging
ACR335355000500 gPlastic Bottle
Catalog number ACR335355000
Price (MYR)
-
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDowex™ 50WX8
Physical FormBeads
CAS11119-67-8
Particle Size15% max. (on 100 mesh, cumulative), 15% max. (through 325 mesh)
PackagingPlastic Bottle